Learn More
Medchemexpress LLC (R,R)-GSK321 | 1816272-19-1 | 99.36% | 100 MG
SDP
Catalog No. 5000338661
Encompass_Preferred
(R,R)-GSK321 is a high-purity wild-type isocitrate dehydrogenase 1 (WT IDH1) inhibitor. This compound acts as an isomer of GSK321, exhibiting potent inhibition with an IC50 value of 120 nM. It also functions as a mutant R132H IDH1 inhibitor with some wild-type cross-reactivity, making it a valuable tool for relevant research.
  • Inhibits wild-type isocitrate dehydrogenase 1 (WT IDH1)
  • IC50 value of 120 nM
  • Functions as a mutant R132H IDH1 inhibitor
  • High purity of 99.36%
  • Appearance as a white to off-white solid
  • For research use only
Catalog No. 50-003-38661 Supplier Medchemexpress LLC Supplier No. HY128888100MG
(R,R)-GSK321 is a high-purity wild-type isocitrate dehydrogenase 1 (WT IDH1) inhibitor. This compound acts as an isomer of GSK321, exhibiting potent inhibition with an IC50 value of 120 nM. It also functions as a mutant R132H IDH1 inhibitor with some wild-type cross-reactivity, making it a valuable tool for relevant research.
  • Inhibits wild-type isocitrate dehydrogenase 1 (WT IDH1)
  • IC50 value of 120 nM
  • Functions as a mutant R132H IDH1 inhibitor
  • High purity of 99.36%
  • Appearance as a white to off-white solid
  • For research use only