Learn More
Medchemexpress LLC (R,R)-GSK321 | 1816272-19-1 | 99.4% | 50 MG
SDP
Catalog No. 5000341001
Encompass_Preferred
(R,R)-GSK321 is a potent inhibitor of wild-type isocitrate dehydrogenase 1 (WT IDH1) with an IC50 value of 120 nM. This compound also acts as a mutant R132H IDH1 inhibitor, being an isomer of GSK321 with some wild-type cross-reactivity.
  • Potent WT IDH1 inhibitor
  • IC50 value of 120 nM for WT IDH1
  • Mutant R132H IDH1 inhibitor
  • Decreases reductive glutaminolysis in A-498 cells
  • For research use only
  • Associated with cancer, cancer targeted therapy, and cancer metabolism research
  • Classified as a metabolic enzyme/protease and isocitrate dehydrogenase (IDH) inhibitor
Catalog No. 50-003-41001 Supplier Medchemexpress LLC Supplier No. HY12888850MG
(R,R)-GSK321 is a potent inhibitor of wild-type isocitrate dehydrogenase 1 (WT IDH1) with an IC50 value of 120 nM. This compound also acts as a mutant R132H IDH1 inhibitor, being an isomer of GSK321 with some wild-type cross-reactivity.
  • Potent WT IDH1 inhibitor
  • IC50 value of 120 nM for WT IDH1
  • Mutant R132H IDH1 inhibitor
  • Decreases reductive glutaminolysis in A-498 cells
  • For research use only
  • Associated with cancer, cancer targeted therapy, and cancer metabolism research
  • Classified as a metabolic enzyme/protease and isocitrate dehydrogenase (IDH) inhibitor