Learn More
Medchemexpress LLC (R,R)-(+)-Hydrobenzoin | 52340-78-0 | 99.8% | C14H14O2 | 10 G
SDP
Catalog No. 5000434048
Encompass_Preferred
(R,R)-(+)-Hydrobenzoin is a chiral secondary alcohol that can be used as a key substrate or a chiral auxiliary reagent in various catalytic reactions and synthetic experiments. It is for research use only.
  • Used as a key substrate or chiral auxiliary reagent.
  • Can serve as a chiral template.
  • Undergoes oxoselenation reaction for asymmetrically functionalizing cyclopentene.
  • Synthesizes chiral cyclopentanol and amino cyclopentanol.
  • Acts as a substrate for deoxygenation and dehydration reactions.
  • Generates trans-stilbene and benzaldehyde with molybdenum (Mo) base catalyst.
Catalog No. 50-004-34048 Supplier Medchemexpress LLC Supplier No. HY5912510G
(R,R)-(+)-Hydrobenzoin is a chiral secondary alcohol that can be used as a key substrate or a chiral auxiliary reagent in various catalytic reactions and synthetic experiments. It is for research use only.
  • Used as a key substrate or chiral auxiliary reagent.
  • Can serve as a chiral template.
  • Undergoes oxoselenation reaction for asymmetrically functionalizing cyclopentene.
  • Synthesizes chiral cyclopentanol and amino cyclopentanol.
  • Acts as a substrate for deoxygenation and dehydration reactions.
  • Generates trans-stilbene and benzaldehyde with molybdenum (Mo) base catalyst.