Learn More
Medchemexpress LLC (R,R)-(+)-Hydrobenzoin | 52340-78-0 | 99.8% | C14H14O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY59125
Encompass_Preferred
(R,R)-(+)-Hydrobenzoin is a chiral secondary alcohol used as a key substrate or chiral auxiliary reagent in various catalytic and synthetic experiments. It can serve as a chiral template in oxoselenation reactions to asymmetrically functionalize cyclopentene, leading to the synthesis of chiral cyclopentanol and aminocyclopentanol. Additionally, it acts as a substrate in deoxygenation and dehydration reactions catalyzed by molybdenum-based catalysts to generate trans-stilbene and benzaldehyde. This product is for research use only.
  • Used as a key substrate or chiral auxiliary reagent
  • Serves as a chiral template in oxoselenation reactions
  • Synthesizes chiral cyclopentanol and aminocyclopentanol
  • Acts as a substrate in deoxygenation and dehydration reactions
  • Generates trans-stilbene and benzaldehyde
  • For research use only
Catalog No. 50-003-58702