Learn More
R&R Lotions R & R LOTION 32 OZ 12/CS
SDP
Catalog No. NC9958605
Encompass
32 oz. with pump I.C. Antistatic Blue hand lotion is enriched with Aloe Vera & Vitamins AD & E. This lotion does not contain any Lanolin, Mineral Oils, Glycerin, or Silicone and will not contaminate print circuit boards. The lotion relieves chapping, skin flaking, irritation and skin sensitivity cau
Catalog No. NC9958605 Supplier R&R Lotions Supplier No. ICL32
32 oz. with pump I.C. Antistatic Blue hand lotion is enriched with Aloe Vera & Vitamins AD & E. This lotion does not contain any Lanolin, Mineral Oils, Glycerin, or Silicone and will not contaminate print circuit boards. The lotion relieves chapping, skin flaking, irritation and skin sensitivity cau