Learn More
Strem, An Ascensus Company CAS# 210057-23-1. 500mg. (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (R,R)-Me-DUPHOS-Rh. MFCD01862466
SDP
Supplier:  Strem, An Ascensus Company 450158500MG
Encompass
CAS# 210057-23-1. 500mg. (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (R,R)-Me-DUPHOS-Rh. MFCD01862466. Molecular Weight: 604.26. Molecular Formula: Rh(C8H12)(C18H28P2)+BF4ae. Color/form: red-orange xtl. Strem# 45-0158. http://www.strem.com/catalog/v/45-0158/
Catalog No. 50-204-5332