Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich (R,R)-(-)-N,N'-Bis(3,5-Di-Tert-Butylsalicylidene)-1,2-Cyclohexanediaminomanganese(III) Chloride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
(R,R)-(-)-N,N'-Bis(3,5-Di-Tert-Butylsalicylidene)-1,2-Cyclohexanediaminomanganese(III) Chloride
Specifications
Specifications
| CAS | 138124-32-0 |
| Melting Point | 330°C to 332°C |
| Molecular Formula | C36H52ClMnN2O2 |
| Linear Formula | [[[(CH3)3C]2C6H2-2-(O-)CH=N]2C6H10]MnCl |
| MDL Number | MFCD02101664 |
| Quantity | 5 g |
| Synonym | (R,R)-Jacobsen's catalyst; Jacobsen's catalyst |
| Molecular Weight (g/mol) | 635.2 g/mol |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'