Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R,R)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine 98.0+%, TCI America™
SDP

Catalog No. D4994200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4994200MG 200 mg
D49941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4994200MG Supplier TCI America Supplier No. D4994200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 415918-91-1
Molecular Formula C37H32Cl2NO2P
Molecular Weight (g/mol) 624.542
MDL Number MFCD08561138
InChI Key PQTVIIOJNJVQOL-JUJAXGASSA-N
Synonym (11bR)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine
PubChem CID 72376409
SMILES CC(C1=CC=CC=C1)N(C(C)C2=CC=CC=C2)P3OC4=C(C5=CC=CC=C5C=C4)C6=C(O3)C=CC7=CC=CC=C76.C(Cl)Cl

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 539.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R,R)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.