Learn More
Medchemexpress LLC HY-N6937 5mg Medchemexpress, (R,R)-Secoisolariciresinol diglucoside CAS:158932-33-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69375MG
Encompass_Preferred
Medchemexpress, HY-N6937 5mg (R,R)-Secoisolariciresinol diglucoside CAS:158932-33-3 (R,R)-Secoisolariciresinol diglucoside ((R,R)-SDG) is the minor isomer of Secoisolariciresinol diglucoside in flaxseed. (R,R)-Secoisolariciresinol diglucoside ((R,R)-SDG) possesses antioxidant and free radical scavenging activities and DNA-radioprotective properties. (R,R)-Secoisolariciresinol diglucoside ((R,R)-SDG) inhibits myeloperoxidase (MPO) activity by suppressing both the peroxidase and chlorination cyclesin inflammatory cells[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0482