Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R,R)-Sinpet.Hbf4

Catalog No. 905542500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-554-2500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-554-2500MG Supplier Sigma Aldrich Supplier No. 905542500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R,R)-SINpEt.HBF4 is a chiral NHC ligand developed in the Glorius Lab for use in Ru(II) catalyzed enantioselective hydrogenations. It has been employed for the regioselective asymmetric hydrogenation of quinoxalines, benzofurans, thiophenes, indolizines, isocoumarins and many other heterocycles.Ligand-Controlled Highly Regioselective and Asymmetric Hydrogenation of Quinoxalines Catalyzed by Ruthenium N-Heterocyclic Carbene ComplexesRuthenium-NHC-Diamine Catalyzed Enantioselective Hydrogenation of Isocoumarins

Specifications

CAS 890152-41-7
Melting Point >300°C
Molecular Formula C18H24Cl2N2Ni
Linear Formula C18H24Cl2N2Ni
Quantity 500 mg
Synonym (4,4'-dtbbpy)NiCl2
Molecular Weight (g/mol) 398 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.