Learn More
Regis Technologies Inc (R,R) Whelk-O 1, Kromasil | 25cm x 4.6mm
SDP
Catalog No. NC1825544
Encompass
The Whelk-O 1 Chiral Stationary Phase is based on 1-(3,5-Dinitrobenzamido)- 1,2,3,4,-tetrahydrophenanthrene. This phase allows separation of racemates from a number of families including amides, epoxides, esters, ureas, carbamates, ethers, aziridines, phosphonates, aldehydes, ketones, carboxylic acids, and alcohols.
Catalog No. NC1825544 Supplier Regis Technologies Inc Supplier No. 1780223300
Add to Cart
Add to Cart
The Whelk-O 1 Chiral Stationary Phase is based on 1-(3,5-Dinitrobenzamido)- 1,2,3,4,-tetrahydrophenanthrene. This phase allows separation of racemates from a number of families including amides, epoxides, esters, ureas, carbamates, ethers, aziridines, phosphonates, aldehydes, ketones, carboxylic acids, and alcohols.