Learn More
CELLTREAT Scientific Products Multiple Cavity Reagent Reservoir, 12 Well, Trough Bottom, PP, Non-sterile, 25/CS
SDP
Catalog No. 501535145
Encompass_Preferred
CELLTREAT Scientific Products Multiple Cavity Reagent Reservoir, 12 Well, Trough Bottom. Reagent Reservoirs are designed to assist in repetitive or robotic liquid transfer. Trough Bottom design promotes funneling of liquid to maximize reagent recovery by pipet tips. Low profile is ideal for small volume robotic tips. Multiple wells allow for dispensing of various reagents. Polypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applications. Storage possible at -80¿C. Autoclavable. Manufactured from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged Non-sterile, 5/bag, 25/case.
Catalog No. 50-153-5145 Supplier CELLTREAT Scientific Products Supplier No. 229562
CELLTREAT Scientific Products Multiple Cavity Reagent Reservoir, 12 Well, Trough Bottom. Reagent Reservoirs are designed to assist in repetitive or robotic liquid transfer. Trough Bottom design promotes funneling of liquid to maximize reagent recovery by pipet tips. Low profile is ideal for small volume robotic tips. Multiple wells allow for dispensing of various reagents. Polypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applications. Storage possible at -80¿C. Autoclavable. Manufactured from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged Non-sterile, 5/bag, 25/case.