Learn More
CELLTREAT Scientific Products 55mL Reagent Reservoir, Polystyrene, White, Individually Wrapped, Sterile, 80/CS
SDP
Catalog No. 501535148
Encompass_Preferred
CELLTREAT Scientific Products 55mL Reagent Reservoir, Individually Wrapped. Reagent Reservoirs are designed to assist in repetitive liquid transfer. White color enhances solution visibility. Angled design promotes funneling of liquid to maximize retrieval. 55mL Reagent Reservoirs have pour spouts on corners and textured gripping areas on the sides. Manufactured from polystyrene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged Sterile, 1/bag, 80/case.
Catalog No. 50-153-5148 Supplier CELLTREAT Scientific Products Supplier No. 229296
CELLTREAT Scientific Products 55mL Reagent Reservoir, Individually Wrapped. Reagent Reservoirs are designed to assist in repetitive liquid transfer. White color enhances solution visibility. Angled design promotes funneling of liquid to maximize retrieval. 55mL Reagent Reservoirs have pour spouts on corners and textured gripping areas on the sides. Manufactured from polystyrene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged Sterile, 1/bag, 80/case.