Learn More
CELLTREAT Scientific Products Single Cavity Reagent Reservoir, 96 Pyramid Bottom, PP, Non-sterile, 25/CS
SDP
Catalog No. 501535146
Encompass_Preferred
CELLTREAT Scientific Products Single Cavity Reagent Reservoir, 96 Pyramid Bottom. Reagent Reservoirs are designed to assist in repetitive or robotic liquid transfer. Pyramid design promotes funneling of liquid to maximize reagent recovery by pipet tips. Low profile is ideal for small volume robotic tips. Polypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applications. Storage possible at -80¿C. Autoclavable. Manufactured from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged Non-sterile, 5/bag, 25/case.
Catalog No. 50-153-5146 Supplier CELLTREAT Scientific Products Supplier No. 229563
CELLTREAT Scientific Products Single Cavity Reagent Reservoir, 96 Pyramid Bottom. Reagent Reservoirs are designed to assist in repetitive or robotic liquid transfer. Pyramid design promotes funneling of liquid to maximize reagent recovery by pipet tips. Low profile is ideal for small volume robotic tips. Polypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applications. Storage possible at -80¿C. Autoclavable. Manufactured from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged Non-sterile, 5/bag, 25/case.