Learn More
Abcam (R)-(-)-Rolipram, Selective PDE4 inhibitor, 50MG
Catalog No. 502707913
Encompass_Preferred
MW 275.34 Da, Purity >98%. Selective PDE4 inhibitor (IC₅₀ = 0.22 μM). More active enantiomer: 2-10-fold more potent that the (S)-(+)-enantiomer (ab120030).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7913 Supplier Abcam Supplier No. AB12003150MG
Add to Cart
Add to Cart
MW 275.34 Da, Purity >98%. Selective PDE4 inhibitor (IC₅₀ = 0.22 μM). More active enantiomer: 2-10-fold more potent that the (S)-(+)-enantiomer (ab120030).
The product is subject to the following: Abcam Restricted Use Statement