Learn More
AdipoGen R-Roscovitine
SDP
Catalog No. 502562400
Encompass_Preferred
Chemical. CAS 186692-46-6. Formula C19H26N6O. MW 354.5. Potent and selective inhibitor of CDKs. More potent than olomoucine. Inhibits CDK1/cyclin B kinase IC50 = 450 nM, CDK2 IC50 = 700 nM and CDK5/p35 IC50 = 160 nM. Inhibits M phase promoting factor MPF kinase activity. Arrests human fibroblasts in G1 phase. Antitumor compound. Activates the mitogen-activated protein kinase pathway. Targets both the p53 and NF-kappaB pathways10. Has effects on calcium channel gating. Prevents DNA damage-induced cyclin A1 upregulation. Apoptosis inducer. As CYC202 in phase I clinical trials. Reviews.
Catalog No. 50-256-2400 Supplier AdipoGen Supplier No. AGCR10006M005
Chemical. CAS 186692-46-6. Formula C19H26N6O. MW 354.5. Potent and selective inhibitor of CDKs. More potent than olomoucine. Inhibits CDK1/cyclin B kinase IC50 = 450 nM, CDK2 IC50 = 700 nM and CDK5/p35 IC50 = 160 nM. Inhibits M phase promoting factor MPF kinase activity. Arrests human fibroblasts in G1 phase. Antitumor compound. Activates the mitogen-activated protein kinase pathway. Targets both the p53 and NF-kappaB pathways10. Has effects on calcium channel gating. Prevents DNA damage-induced cyclin A1 upregulation. Apoptosis inducer. As CYC202 in phase I clinical trials. Reviews.