Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-RUCY(regR)-XylBINAP, TCI America™
SDP

Catalog No. R0139200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0139200MG 200 mg
R01391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0139200MG Supplier TCI America Supplier No. R0139200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1384974-38-2
Molecular Formula C71H73ClN2O2P2Ru
MDL Number MFCD20261161
Synonym RuCl[(R)-daipena][(R)-xylbinap]

Specifications

Color Green-Yellow
Quantity 200 mg
UN Number 3077
Formula Weight 1184.84
Physical Form Crystalline Powder
Chemical Name or Material (R)-RUCY(regR)-XylBINAP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.