Learn More
Strem, An Ascensus Company CAS# 1384974-38-2. 250mg. Chloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2R)-(-)-1-(4-methoxyphenyl)-1’-(4-methoxyphenyl-kC)-3-methyl-1,2-butanediamine]ruthenium(II) (R)-RUCY-XylBINAP. MFCD20261161
SDP
Supplier:  Strem, An Ascensus Company 440217250MG
Encompass
CAS# 1384974-38-2. 250mg. Chloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2R)-(-)-1-(4-methoxyphenyl)-1’-(4-methoxyphenyl-kC)-3-methyl-1,2-butanediamine]ruthenium(II) (R)-RUCY-XylBINAP. MFCD20261161. Molecular Weight: 1184.82. Molecular Formula: C71H73ClN2O2P2Ru. Color/form: yellow to dark brown/green solid Strem# 44-0217. http://www.strem.com/catalog/v/44-0217/
Catalog No. 50-201-6718