Learn More
Cayman Chemical R S-3 5-DHPG 50mg
SDP
Catalog No. 502912502
Encompass_Preferred
An agonist of group I mGluRs; induces phosphoinositol hydrolysis in MCB3901 cells expressing human mGluR1a and mGluR5a; potentiates NMDA-induced depolarizations in rat hippocampal slices at 10 µM; intrathecal administration increases mean blood pressure in anesthetized rats at 300 nmol/animal; induces seizures in rats at 1.5 µmol/animal, i.c.v.
Catalog No. 50-291-2502 Supplier Cayman Chemical Supplier No. 3591250 MG
An agonist of group I mGluRs; induces phosphoinositol hydrolysis in MCB3901 cells expressing human mGluR1a and mGluR5a; potentiates NMDA-induced depolarizations in rat hippocampal slices at 10 µM; intrathecal administration increases mean blood pressure in anesthetized rats at 300 nmol/animal; induces seizures in rats at 1.5 µmol/animal, i.c.v.