Learn More
Cayman Chemical (R,S)-AMPA (hydrate), Purity: ≥95%, 50 mg
SDP
Catalog No. 502971894
Encompass_Preferred
The defining agonist for the AMPA subgroup of ionotropic glutamate receptors, which mediate excitatory neurotransmission; contains an equal mixture of the active (S)-enantiomer and the inactive (R)-enantiomer
Catalog No. 50-297-1894 Supplier Cayman Chemical Supplier No. 1457150 MG
The defining agonist for the AMPA subgroup of ionotropic glutamate receptors, which mediate excitatory neurotransmission; contains an equal mixture of the active (S)-enantiomer and the inactive (R)-enantiomer