Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R,S)-anatabine | 2743-90-0 | 99.7% | 160.22 g/mol | C10H12N2 | 25MG
SDP

Catalog No. 5000235544
Encompass_Preferred

(R,S)-Anatabine is a minor tobacco alkaloid used as an analytical standard and as a specific marker for detection of tobacco use. It is supplied for research and analytical applications in high-purity solid and solution formats. Molecular formula C10H12N2; molecular weight 160.22 g/mol.

  • High purity (99.7%).
  • Chemical formula C10H12N2.
  • Molecular weight 160.22 g/mol.
  • Available as solid quantities (5 mg-100 mg) and as a 10 mM solution in DMSO.
  • Intended for analytical and biochemical research applications, including tobacco use detection.

Catalog No. 50-002-35544 Supplier Medchemexpress LLC Supplier No. HY126047A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R,S)-Anatabine is a minor tobacco alkaloid used as an analytical standard and as a specific marker for detection of tobacco use. It is supplied for research and analytical applications in high-purity solid and solution formats. Molecular formula C10H12N2; molecular weight 160.22 g/mol.

  • High purity (99.7%).
  • Chemical formula C10H12N2.
  • Molecular weight 160.22 g/mol.
  • Available as solid quantities (5 mg-100 mg) and as a 10 mM solution in DMSO.
  • Intended for analytical and biochemical research applications, including tobacco use detection.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.