Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R,S-anatabine | 2743-90-0 | MFCD01669938 | 99.7% | 160.22 | C10H12N2 | 100MG
SDP

Catalog No. 5000236437
Encompass_Preferred

(R,S)-Anatabine is the racemic form of anatabine, a minor tobacco alkaloid supplied as an analytical standard and research reagent for biochemical and analytical applications, including use as a marker of tobacco exposure and in amyloid-beta studies.

  • Analytical standard for detection and quantification of tobacco alkaloids.
  • Racemic (R,S) form of anatabine.
  • Molecular formula C10H12N2; molecular weight 160.22.
  • High purity (99.7%).
  • Typically supplied as a 100 mg reference material.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-36437 Supplier Medchemexpress LLC Supplier No. HY126047A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R,S)-Anatabine is the racemic form of anatabine, a minor tobacco alkaloid supplied as an analytical standard and research reagent for biochemical and analytical applications, including use as a marker of tobacco exposure and in amyloid-beta studies.

  • Analytical standard for detection and quantification of tobacco alkaloids.
  • Racemic (R,S) form of anatabine.
  • Molecular formula C10H12N2; molecular weight 160.22.
  • High purity (99.7%).
  • Typically supplied as a 100 mg reference material.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.