Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R,S)-Fmoc-3-amino-3-(biphenyl)-propionic acid

Catalog No. CDS004401 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS004401 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS004401 Supplier Sigma Aldrich Supplier No. CDS004401100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The isothermal vapor-liquid equilibrium for propionic acid and butyl propionate was studied
  • The rate co-efficient for the gas-phase reaction of butyl propionate with oh radical was studied by both absolute and relative methods

Specifications

Molecular Formula C30H25NO4
Linear Formula C30H25NO4
Quantity 250 mL
MDL Number MFCD02179646
Molecular Weight (g/mol) 463.52
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.