Learn More
Cayman Chemical (R,S)-N-Nitrosoanatabine, Purity: ≥98%, CAS: 887407-16-1, 5 mg
SDP
Catalog No. 502949823
Encompass_Preferred
A tobacco-specific nitrosamine; not carcinogenic in rats at 1, 3, and 9 mmol/kg
Catalog No. 50-294-9823 Supplier Cayman Chemical Supplier No. 338515 MG
A tobacco-specific nitrosamine; not carcinogenic in rats at 1, 3, and 9 mmol/kg