Learn More
Medchemexpress LLC (R,s)-((oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) bis(2-(4-((4s,5r)-4,5-bis(4-chlorophen | 2249750-23-8 | 95.0% | 1437.25 g/mol | C72H78Cl4N8O15 | 5 MG
SDP
Catalog No. 5000262481
Encompass_Preferred
PROTAC MDM2 Degrader-3 is a proteolysis-targeting chimera (PROTAC) designed to induce selective degradation of MDM2 by recruiting an E3 ubiquitin ligase, enabling proteasomal removal of the target protein. It is formulated as a solid research reagent intended for in vitro studies and is supplied with batch-specific analytical documentation.
Catalog No. 50-002-62481 Supplier Medchemexpress LLC Supplier No. HY1288425MG
PROTAC MDM2 Degrader-3 is a proteolysis-targeting chimera (PROTAC) designed to induce selective degradation of MDM2 by recruiting an E3 ubiquitin ligase, enabling proteasomal removal of the target protein. It is formulated as a solid research reagent intended for in vitro studies and is supplied with batch-specific analytical documentation.