Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R,S,S,R,S)-Boc-Dap-NE | 2994330-72-0 | 97.1% | 420.54 | C23H36N2O5 | 10 MG
SDP

Catalog No. 5000298504
Encompass_Preferred

(R,S,S,R,S)-Boc-Dap-NE is the inactive stereoisomer of Boc-Dap-NE supplied as a research chemical and synthetic intermediate. It is intended for use as an experimental control and as a building-block intermediate in the synthesis of monomethyl auristatin E; safety documentation and analytical data are provided for research use.

  • Inactive stereoisomer suitable as experimental control.
  • Serves as intermediate in monomethyl auristatin E synthesis.
  • Supplied in a 10 MG quantity for laboratory use.
  • Reported purity approximately 97.1%.
  • Molecular weight 420.54 g·mol⁻¹; formula C23H36N2O5.

Catalog No. 50-002-98504 Supplier Medchemexpress LLC Supplier No. HY78931A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R,S,S,R,S)-Boc-Dap-NE is the inactive stereoisomer of Boc-Dap-NE supplied as a research chemical and synthetic intermediate. It is intended for use as an experimental control and as a building-block intermediate in the synthesis of monomethyl auristatin E; safety documentation and analytical data are provided for research use.

  • Inactive stereoisomer suitable as experimental control.
  • Serves as intermediate in monomethyl auristatin E synthesis.
  • Supplied in a 10 MG quantity for laboratory use.
  • Reported purity approximately 97.1%.
  • Molecular weight 420.54 g·mol⁻¹; formula C23H36N2O5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.