Learn More
Lkt Laboratories R,S-Sulforaphane, ≥99%, 25MG
SDP
Catalog No. 501534837
Encompass_Preferred
Phase II enzyme inducer; Nrf2 activator; antioxidant with anticarcinogenic properties. CAS: 4478-93-7. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4837 Supplier Lkt Laboratories Supplier No. S804425MG
Phase II enzyme inducer; Nrf2 activator; antioxidant with anticarcinogenic properties. CAS: 4478-93-7. Additional sizes available upon request. Ships from Saint Paul, MN.