Learn More
Seracare Life Sciences Inc R SALMONINARUM POS CTR
Supplier:  Seracare Life Sciences Inc 53700022
Encompass_Preferred
Heat-killed Renibacterium salmoninarum cells. It is intended for use as a positive control in immunoassays designed for R. salmoninarum detection.
Catalog No. 50-674-68