Learn More
AdipoGen R-Semivioxanthin
SDP
Supplier:  AdipoGen BVT0360C500
Encompass_Preferred
Chemical. CAS 70477-26-8. Formula C15H14O5. MW 274.3. Isolated from Penicillium citreo-viride. Mycotoxin. Antibacterial Gram-positive and antifungal. Immunoregulator by degradation of IkappaB Inhibitor of NF-kappaB and regulation of the TNF-alpha tumor necrosis factor-alpha and MAPK mitogen-activated protein kinase signaling pathways.
Catalog No. 50-256-1974