Learn More
AdipoGen R-Semixanthomegnin
SDP
Supplier:  AdipoGen BVT0366M001
Encompass_Preferred
Chemical. Formula C15H12O6. MW 288.3. Derivative of semivioxanthin from Penicillium citreo-viride. Antibacterial mycotoxin derivative. Monomer half of the genotoxic mycotoxin xanthomegnin.
Catalog No. 50-256-1967