Learn More
Medchemexpress LLC HY-100808 1g Medchemexpress, (R)-Serine CAS:312-84-5 Purity:>98%
SDP
Catalog No. 501931956
Encompass_Preferred
Medchemexpress, HY-100808 1g (R)-Serine CAS:312-84-5 (R)-Serine, an endogenous amino acid involved in glia-synapse interactions that has unique neurotransmitter characteristics, is a potent co-agonist at the NMDA glutamate receptor . (R)-Serine has a cardinal modulatory role in major NMDAR-dependent processes including NMDAR-mediated neurotransmission, neurotoxicity, synaptic plasticity, and cell migration [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1956 Supplier Medchemexpress LLC Supplier No. HY1008081G
Medchemexpress, HY-100808 1g (R)-Serine CAS:312-84-5 (R)-Serine, an endogenous amino acid involved in glia-synapse interactions that has unique neurotransmitter characteristics, is a potent co-agonist at the NMDA glutamate receptor . (R)-Serine has a cardinal modulatory role in major NMDAR-dependent processes including NMDAR-mediated neurotransmission, neurotoxicity, synaptic plasticity, and cell migration [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.