Learn More
Cayman Chemical R Shydroxy Ramelton Meta 10mg
SDP
Catalog No. 502947826
Encompass_Preferred
A mixed isomer preparation of ramelteon metabolite M-II; has an (R,S) configuration at the hydroxy group, where ramelteon metabolite M-II has the (S) configuration at that position
Catalog No. 50-294-7826 Supplier Cayman Chemical Supplier No. 3389710 MG
A mixed isomer preparation of ramelteon metabolite M-II; has an (R,S) configuration at the hydroxy group, where ramelteon metabolite M-II has the (S) configuration at that position