Learn More
Medchemexpress LLC HY-100888A 2mg Medchemexpress, (R)-Simurosertib CAS:1330782-69-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100888A2MG
Encompass_Preferred
Medchemexpress, HY-100888A 2mg (R)-Simurosertib CAS:1330782-69-8 (R)-Simurosertib ((R)-TAK-931) is the (R)-enantiomer of Simurosertib, Simurosertib (TAK-931) is a selective cycle 7 (CDC7) kinase inhibitor, with an IC50<0.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7787