Learn More
Medchemexpress LLC HY-100888A 5mg Medchemexpress, (R)-Simurosertib CAS:1330782-69-8 Purity:>98%
SDP
Catalog No. 501931989
Encompass_Preferred
Medchemexpress, HY-100888A 5mg (R)-Simurosertib CAS:1330782-69-8 (R)-Simurosertib ((R)-TAK-931) is the (R)-enantiomer of Simurosertib, Simurosertib (TAK-931) is a selective cycle 7 (CDC7) kinase inhibitor, with an IC 50 <0.3 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1989 Supplier Medchemexpress LLC Supplier No. HY100888A5MG
Medchemexpress, HY-100888A 5mg (R)-Simurosertib CAS:1330782-69-8 (R)-Simurosertib ((R)-TAK-931) is the (R)-enantiomer of Simurosertib, Simurosertib (TAK-931) is a selective cycle 7 (CDC7) kinase inhibitor, with an IC 50 <0.3 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.