Learn More
Abcam (R)-(+)-SKF38393 hydrochloride, D1 partial agonist, 5MG
Catalog No. 502707415
Encompass_Preferred
MW 291.77 Da, Purity >98%. Selective D1 receptor partial agonist. Active enantiomer of (±)-SKF-38393 (ab120740).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7415 Supplier Abcam Supplier No. AB1206195MG
MW 291.77 Da, Purity >98%. Selective D1 receptor partial agonist. Active enantiomer of (±)-SKF-38393 (ab120740).
The product is subject to the following: Abcam Restricted Use Statement