Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R-spondin-2 (RSPO2) protein, mouse, Fc-fusion | >85.0% | 50 UG
SDP

Catalog No. 5000292010
Encompass_Preferred

Recombinant mouse R-spondin-2 Fc-fusion protein produced in HEK293 cells, supplied as a purified, sterile-filtered aqueous solution for research use. The protein comprises the extracellular region (N24-G205) fused to a C-terminal Fc tag and is intended for signaling and cell-culture assays.

  • Species: mouse
  • Expression system: HEK293 mammalian cells
  • Construct: N24-G205, C-terminal Fc fusion
  • Predicted molecular weight: ~47.4 kDa (observed ~45-50 kDa)
  • Purity: >85.0%
  • Endotoxin: <1 EU/μg
  • Formulation: PBS, pH 7.4, 0.22 μm filtered
  • Storage: store at -80°C; avoid repeated freeze-thaw cycles
  • Intended use: for research use only

Catalog No. 50-002-92010 Supplier Medchemexpress LLC Supplier No. HYP7601350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse R-spondin-2 Fc-fusion protein produced in HEK293 cells, supplied as a purified, sterile-filtered aqueous solution for research use. The protein comprises the extracellular region (N24-G205) fused to a C-terminal Fc tag and is intended for signaling and cell-culture assays.

  • Species: mouse
  • Expression system: HEK293 mammalian cells
  • Construct: N24-G205, C-terminal Fc fusion
  • Predicted molecular weight: ~47.4 kDa (observed ~45-50 kDa)
  • Purity: >85.0%
  • Endotoxin: <1 EU/μg
  • Formulation: PBS, pH 7.4, 0.22 μm filtered
  • Storage: store at -80°C; avoid repeated freeze-thaw cycles
  • Intended use: for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.