Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-STU104-d6 | 3063042-81-6 | 99.9% | 304.37 g/mol | C18H12D6O4 | 10 MG
SDP

Catalog No. 5000286438
Encompass_Preferred

(R)-STU104-d6 is the deuterium-labeled (d6) analog of (R)-STU104, a TAK1-MKK3 protein-protein interaction inhibitor. Supplied as a 10 mg solid research reagent (CAS 3063042-81-6), it has molecular formula C18H12D6O4 and molecular weight 304.37 g/mol. The material is characterized for analytical use and supplied with supporting documentation.

  • Deuterium-labeled (d6) analog suitable as an isotopic tracer or internal standard.
  • Targets TAK1-MKK3 protein-protein interaction for mechanistic studies.
  • High assay purity (reported 99.9% by LCMS).
  • Molecular formula C18H12D6O4 and molecular weight 304.37 g/mol.
  • Supplied as a 10 mg solid, convenient for analytical and small-scale studies.
  • Provided with certificate of analysis and safety data sheet for traceability.

Catalog No. 50-002-86438 Supplier Medchemexpress LLC Supplier No. HY150612S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-STU104-d6 is the deuterium-labeled (d6) analog of (R)-STU104, a TAK1-MKK3 protein-protein interaction inhibitor. Supplied as a 10 mg solid research reagent (CAS 3063042-81-6), it has molecular formula C18H12D6O4 and molecular weight 304.37 g/mol. The material is characterized for analytical use and supplied with supporting documentation.

  • Deuterium-labeled (d6) analog suitable as an isotopic tracer or internal standard.
  • Targets TAK1-MKK3 protein-protein interaction for mechanistic studies.
  • High assay purity (reported 99.9% by LCMS).
  • Molecular formula C18H12D6O4 and molecular weight 304.37 g/mol.
  • Supplied as a 10 mg solid, convenient for analytical and small-scale studies.
  • Provided with certificate of analysis and safety data sheet for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.