Learn More
Medchemexpress LLC (R)-STU104-d6 | 3063042-81-6 | C18H12D6O4 | 1 MG
SDP
Catalog No. 5000375101
Encompass_Preferred
R-STU104-d6 is a chemical substance supplied as a solid. It is intended for laboratory chemical use and the manufacture of substances. This product is stable under recommended storage conditions and should be handled with appropriate personal protective equipment and ventilation.
  • Stable under recommended storage conditions
  • Suitable for laboratory chemical applications
  • Supplied as a solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Can be shipped at room temperature if less than 2 weeks
Catalog No. 50-003-75101 Supplier Medchemexpress LLC Supplier No. HY150612S1MG
R-STU104-d6 is a chemical substance supplied as a solid. It is intended for laboratory chemical use and the manufacture of substances. This product is stable under recommended storage conditions and should be handled with appropriate personal protective equipment and ventilation.
  • Stable under recommended storage conditions
  • Suitable for laboratory chemical applications
  • Supplied as a solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Can be shipped at room temperature if less than 2 weeks