Learn More
Enzo Life Sciences R-Sulforaphane (10mg). CAS: 142825-10-3
SDP
Catalog No. 502008789
Encompass_Preferred
Potent, selective inducer of phase II detoxification enzymes. Inhibits chemically induced mammary tumor formation in rats. Alternative name: L-Sulforaphane, (-)-1-Isothiocyanato-4R-(methylsulfinyl)-butane. Purity: ≥98% (UHPLC). Solubility: Soluble in methanol, 100% ethanol, DMSO or ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-200-8789 Supplier Enzo Life Sciences Supplier No. ALX350230M010
Potent, selective inducer of phase II detoxification enzymes. Inhibits chemically induced mammary tumor formation in rats. Alternative name: L-Sulforaphane, (-)-1-Isothiocyanato-4R-(methylsulfinyl)-butane. Purity: ≥98% (UHPLC). Solubility: Soluble in methanol, 100% ethanol, DMSO or ethyl acetate. Long Term Storage: -20°C.