Learn More
Medchemexpress LLC R-sulforaphane | 142825-10-3 | >98.0% | C6H11NOS2 | 10MG
SDP
Catalog No. 5000258486
Encompass_Preferred
(R)-Sulforaphane is the R-enantiomer of sulforaphane, an organosulfur isothiocyanate (CAS 142825-10-3) that induces phase II detoxification enzymes and activates the Nrf2 antioxidant pathway. It is used in biochemical and cell-based research to study antioxidant response, detoxification, inflammation, and cancer-related signaling. Supplied as a purified research-grade compound (purity >98%).
  • Chiral R-enantiomer with defined stereochemistry.
  • Activates Nrf2-mediated antioxidant response.
  • Induces phase II detoxification enzymes.
  • Suitable for cell-based and biochemical assays.
  • High purity for reproducible results.
  • Small, well-characterized organic molecule (C6H11NOS2, 177.29 g/mol).
Catalog No. 50-002-58486 Supplier Medchemexpress LLC Supplier No. HY13755A10MG
(R)-Sulforaphane is the R-enantiomer of sulforaphane, an organosulfur isothiocyanate (CAS 142825-10-3) that induces phase II detoxification enzymes and activates the Nrf2 antioxidant pathway. It is used in biochemical and cell-based research to study antioxidant response, detoxification, inflammation, and cancer-related signaling. Supplied as a purified research-grade compound (purity >98%).
  • Chiral R-enantiomer with defined stereochemistry.
  • Activates Nrf2-mediated antioxidant response.
  • Induces phase II detoxification enzymes.
  • Suitable for cell-based and biochemical assays.
  • High purity for reproducible results.
  • Small, well-characterized organic molecule (C6H11NOS2, 177.29 g/mol).