Learn More
TARGETMOL CHEMICALS INC R-Sulforaphane 5MG
SDP
Catalog No. 502876987
Encompass_Preferred
Also available in 1 mg, 2 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (R)-Sulforaphane (L-Sulforaphane) is a potent inducer of the Keap1/Nrf2/ARE pathway.
Catalog No. 50-287-6987 Supplier TARGETMOL CHEMICALS INC Supplier No. T82815MG
Also available in 1 mg, 2 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (R)-Sulforaphane (L-Sulforaphane) is a potent inducer of the Keap1/Nrf2/ARE pathway.