Learn More
Vector Laboratories Animal-Free Blocker and Diluent (AFB), Ready-to-Use (R.T.U.) 100 ml
SDP
Catalog No. NC1404986
Encompass
R.T.U. Animal-Free Blocker and Diluent is a ready-to-use antibody diluent and blocking reagent that stabilizes diluted antibody solutions and diminishes background due to non-specific binding in immunohistochemical applications. It contains no animal-derived proteins and can be used as an alternative to sera, BSA, casein or non-fat dry milk in situations where these or other traditional blocking agents are ineffective. It is especially convenient for multiplex IHC (multiple antigen labeling) applications where antibodies from several different species are used.
Catalog No. NC1404986 Supplier Vector Laboratories Supplier No. SP5035100
R.T.U. Animal-Free Blocker and Diluent is a ready-to-use antibody diluent and blocking reagent that stabilizes diluted antibody solutions and diminishes background due to non-specific binding in immunohistochemical applications. It contains no animal-derived proteins and can be used as an alternative to sera, BSA, casein or non-fat dry milk in situations where these or other traditional blocking agents are ineffective. It is especially convenient for multiplex IHC (multiple antigen labeling) applications where antibodies from several different species are used.