Learn More
Vector Laboratories Goat Anti-Rabbit IgG Antibody Biotinylated Ready to Use 50 ml
SDP
Supplier:  Vector Laboratories BP910050
Encompass
This stabilized, prediluted, ready-to-use (R.T.U.) solution of biotinylated goat anti-rabbit secondary antibody is provided in a convenient dropper bottle format. The high-affinity antibody is adsorbed against serum and immunoglobulins from potentially interfering species and biotinylated in a manner that ensures the maximum degree of labeling without compromising the specificity or affinity of the antibody. It can be used for tissue and cell staining, ELISAs, and blots.
Catalog No. NC1458691