Learn More
Medchemexpress LLC (R)-Terazosin | 109351-34-0 | 5 MG
SDP
Catalog No. 5000405964
Encompass_Preferred
(R)-Terazosin is an active R-enantiomer of Terazosin and a potent α1-adrenoceptor antagonist. It exhibits Ki values of 6.51 nM, 1.01 nM, and 1.97 nM for α1a, α1b, and α1d-adrenoceptor, respectively.
  • Potent α1-adrenoceptor antagonist
  • Low affinity for α2a, α2B, and α2c-adrenoceptors
  • Useful probe in understanding the functional role of adrenoceptor subtypes
  • Weaker antagonist at α2B sites than its enantiomer for differentiating pharmacological effects
  • Powder stable for 3 years at -20°C and 2 years at 4°C
  • In solvent, stable for 2 years at -80°C and 1 year at -20°C
  • Soluble in DMSO
Catalog No. 50-004-05964 Supplier Medchemexpress LLC Supplier No. HYB0371B5MG
(R)-Terazosin is an active R-enantiomer of Terazosin and a potent α1-adrenoceptor antagonist. It exhibits Ki values of 6.51 nM, 1.01 nM, and 1.97 nM for α1a, α1b, and α1d-adrenoceptor, respectively.
  • Potent α1-adrenoceptor antagonist
  • Low affinity for α2a, α2B, and α2c-adrenoceptors
  • Useful probe in understanding the functional role of adrenoceptor subtypes
  • Weaker antagonist at α2B sites than its enantiomer for differentiating pharmacological effects
  • Powder stable for 3 years at -20°C and 2 years at 4°C
  • In solvent, stable for 2 years at -80°C and 1 year at -20°C
  • Soluble in DMSO