Learn More
Medchemexpress LLC (R)-Terazosin | 109351-34-0 | 99.8% | 100 MG
SDP
Catalog No. 5000405473
Encompass_Preferred
(R)-Terazosin is an active R-enantiomer of Terazosin and a potent α1-adrenoceptor antagonist. It exhibits Ki values of 6.51 nM, 1.01 nM, and 1.97 nM for α1a, α1b, and α1d-adrenoceptor, respectively.
  • Potent α1-adrenoceptor antagonist
  • Low affinity for α2a, α2B, and α2c-adrenoceptor
  • Useful probe for understanding functional roles of adrenoceptor subtypes
  • Antagonism at rat atrial α2B receptor
  • Antagonism at rat vas deferens α1A and α2A receptor
Catalog No. 50-004-05473 Supplier Medchemexpress LLC Supplier No. HYB0371B100MG
(R)-Terazosin is an active R-enantiomer of Terazosin and a potent α1-adrenoceptor antagonist. It exhibits Ki values of 6.51 nM, 1.01 nM, and 1.97 nM for α1a, α1b, and α1d-adrenoceptor, respectively.
  • Potent α1-adrenoceptor antagonist
  • Low affinity for α2a, α2B, and α2c-adrenoceptor
  • Useful probe for understanding functional roles of adrenoceptor subtypes
  • Antagonism at rat atrial α2B receptor
  • Antagonism at rat vas deferens α1A and α2A receptor