Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-terazosin | 109351-34-0 | 99.8% | 387.43 g·mol⁻1 | C19H25N5O4 | 10 MG
SDP

Catalog No. 5000298216
Encompass_Preferred

(R)-Terazosin is the active R-enantiomer of terazosin, a potent α1-adrenoceptor antagonist supplied as a solid research chemical for in vitro and preclinical pharmacology studies. It is used in receptor pharmacology assays and studies of α1-adrenergic signaling.

  • Potent α1-adrenoceptor antagonist, active R-enantiomer.
  • CAS number 109351-34-0 for unambiguous identification.
  • High purity (approximately 99.8%) suitable for research use.
  • Molecular formula C19H25N5O4; molecular weight 387.43 g·mol⁻1.
  • Solid appearance; soluble in DMSO with ready-made 10 mM solutions available.
  • Provided in small research pack sizes appropriate for assay development (10 mg format).

Catalog No. 50-002-98216 Supplier Medchemexpress LLC Supplier No. HYB0371B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Terazosin is the active R-enantiomer of terazosin, a potent α1-adrenoceptor antagonist supplied as a solid research chemical for in vitro and preclinical pharmacology studies. It is used in receptor pharmacology assays and studies of α1-adrenergic signaling.

  • Potent α1-adrenoceptor antagonist, active R-enantiomer.
  • CAS number 109351-34-0 for unambiguous identification.
  • High purity (approximately 99.8%) suitable for research use.
  • Molecular formula C19H25N5O4; molecular weight 387.43 g·mol⁻1.
  • Solid appearance; soluble in DMSO with ready-made 10 mM solutions available.
  • Provided in small research pack sizes appropriate for assay development (10 mg format).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.