Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-tert-Leucinol, 98%

Catalog No. AC436671000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC436671000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436671000 Supplier Thermo Scientific Chemicals Supplier No. 436671000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 112245-09-7 | C6H15NO | 117.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112245-09-7
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD01076574
InChI Key JBULSURVMXPBNA-MLHKIVSYNA-N
Synonym r-tert-leucinol, d-tert-leucinol, 2r-2-amino-3,3-dimethylbutan-1-ol, r-2-amino-3,3-dimethylbutan-1-ol, r-2-amino-3,3-dimethyl-1-butanol, 2r-2-amino-3,3-dimethyl-1-butanol, r---tert-leucinol, ?-tert-leucinol, r-?-tert-leucinol
PubChem CID 6932847
IUPAC Name (2R)-2-amino-3,3-dimethylbutan-1-ol
SMILES CC(C)(C)[C@@H](N)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation -34° to -40° (20°C, 589nm) (c=0.7, ethanol)
Density 0.9g/mL
Quantity 100 mg
Formula Weight 117.19
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material (R)-(-)-tert-Leucinol
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.