Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Tetrahydrofuran-2-yl-methylamine | 7202-43-9 | ≥97.0% | 101.15 | 250 MG
SDP

Catalog No. 5000453277
Encompass_Preferred

(R)-Tetrahydrofuran-2-yl-methylamine (1-[(2R)-tetrahydrofuran-2-yl]methanamine) is a synthetic intermediate useful for pharmaceutical synthesis. It is for research use only.

  • Molecular weight: 101.15
  • CAS No.: 7202-43-9
  • Formula: C5H11NO
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Purity: ≥97.0%

Catalog No. 50-004-53277 Supplier Medchemexpress LLC Supplier No. 250MGHYW010601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Tetrahydrofuran-2-yl-methylamine (1-[(2R)-tetrahydrofuran-2-yl]methanamine) is a synthetic intermediate useful for pharmaceutical synthesis. It is for research use only.

  • Molecular weight: 101.15
  • CAS No.: 7202-43-9
  • Formula: C5H11NO
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Purity: ≥97.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.