Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-Tetrahydrofurfurylamine 98.0+%, TCI America™
SDP

Catalog No. T2551200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2551200MG 200 mg
T25511G 1 g
T25515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T2551200MG Supplier TCI America Supplier No. T2551200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7202-43-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00192476
InChI Key YNOGYQAEJGADFJ-RXMQYKEDSA-N
Synonym r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2-furanmethanamine, tetrahydro-, 2r, 1-2r-tetrahydrofuran-2-yl methanamine, pubchem7001, 2r-tetrahydrofurfurylamine
PubChem CID 2734124
IUPAC Name [(2R)-oxolan-2-yl]methanamine
SMILES C1CC(OC1)CN

Specifications

Color Yellow
Quantity 200 mg
Physical Form Clear Liquid at 20°C
UN Number 2943
Boiling Point 60°C
Formula Weight 101.15
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (R)-(-)-Tetrahydrofurfurylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.