Learn More
Medchemexpress LLC (R)-(+)-thalidomide | 260939 | MFCD00210220 | 99.8% | C13H10N2O4 | 10MG
SDP
Catalog No. 5000258734
Encompass_Preferred
(R)-Thalidomide is the R-enantiomer of thalidomide supplied for research use. It is reported to have psychomotor stabilizing properties and is used as a ligand in studies of cereblon-mediated E3 ligase interactions and related cellular pathways. Provided as a high-purity research chemical for biochemical and cellular assays.
  • R-enantiomer of thalidomide for stereospecific studies.
  • High purity 99.8% suitable for analytical and biological assays.
  • Useful for research on cereblon and E3 ligase interactions.
  • Available in milligram scales for laboratory research.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-58734 Supplier Medchemexpress LLC Supplier No. HY14658B10MG
(R)-Thalidomide is the R-enantiomer of thalidomide supplied for research use. It is reported to have psychomotor stabilizing properties and is used as a ligand in studies of cereblon-mediated E3 ligase interactions and related cellular pathways. Provided as a high-purity research chemical for biochemical and cellular assays.
  • R-enantiomer of thalidomide for stereospecific studies.
  • High purity 99.8% suitable for analytical and biological assays.
  • Useful for research on cereblon and E3 ligase interactions.
  • Available in milligram scales for laboratory research.
  • For research use only; not for human or clinical use.